Cyclohexyldiphenylphosphine ≥97.0% (por GC)

Proveedor: TCI
Atención

C1429-25G C1429-5G
TCIAC1429-25GEA 167 EUR
TCIAC1429-25G TCIAC1429-5G
Cyclohexyldiphenylphosphine ≥97.0% (por GC)
Cyclohexyldiphenylphosphine
Formula: C₁₈H₂₁P
Número MDL: MFCD00046360
Núm. CAS: 6372-42-5
EINECS: 228-904-2

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 59.0 to 62.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR