4-Methyl-2-(4-pyridinyl)-5-thiazolecarboxylic acid

Proveedor: Cayman Chemical

Sinónimos: 4-Methyl-2-(pyridin-4-yl)thiazole-5-carboxylic acid

13049-100 13049-250 13049-500 13049-1
CAYM13049-100EA 24.2 EUR
CAYM13049-100 CAYM13049-250 CAYM13049-500 CAYM13049-1
4-Methyl-2-(4-pyridinyl)-5-thiazolecarboxylic acid
4-Methyl-2-(4-pyridinyl)-5-thiazolecarboxylic acid
Formula: C₁₀H₈N₂O₂S
MW: 220,2477 g/mol
Número MDL: MFCD00171757
Núm. CAS: 144060-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR