1,1'-Carbonyldi(1H-1,2,4-triazole) ≥97.0% (by titrimetric analysis)
Proveedor: TCI
|
Peligro
|
C2325-25G
C2325-5G
TCIAC2325-25GEA
364
EUR
TCIAC2325-25G
TCIAC2325-5G
1,1'-Carbonyldi(1H-1,2,4-triazole) ≥97.0% (by titrimetric analysis)
1,1'-Carbonyldi(1H-1,2,4-triazole)
Formula:
C₅H₄N₆O Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00043399 Núm. CAS: 41864-22-6 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 148.0 to 152.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...