1-Hydroxy-4-(p-toluidino)-9,10-anthraquinone ≥90.0% (by HPLC)

Proveedor: TCI

Sinónimos: 1-Hydroxy-4-(p-tolylamino)anthracene-9,10-dione

Q0022-25G Q0022-5G
TCIAQ0022-25GEA 125 EUR
TCIAQ0022-25G TCIAQ0022-5G
1-Hydroxy-4-(p-toluidino)-9,10-anthraquinone ≥90.0% (by HPLC)
1-Hydroxy-4-(p-toluidino)-9,10-anthraquinone
Formula: C₂₁H₁₅NO₃
Número MDL: MFCD00019148
Núm. CAS: 81-48-1
EINECS: 201-353-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94d47dcac47307d57a694704cce9883c8355b364&vid=94d47dcac47307d57a694704cce9883c8355b364&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Red to Dark blue to Black powder to crystal
Purity (HPLC) min. 90.0 area%
Lambda max. 581.0 to 585.0 nm (Toluene)
Absorbance (E1%1cm) min. 310 (Toluene, 581.0 to 585.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR