4-Iodo-1H-pyrazole ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-Iodopyrazole
I0692-25G
I0692-5G
TCIAI0692-25GEA
235
EUR
TCIAI0692-25G
TCIAI0692-5G
4-Iodo-1H-pyrazole ≥98.0% (by GC, titration analysis)
4-Iodo-1H-pyrazole
Formula:
C₃H₃IN₂ MW: 193,97 g/mol |
Número MDL:
MFCD00005244 Núm. CAS: 3469-69-0 EINECS: 222-434-1 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 107.0 to 111.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...