3-Amino-5-hydroxy-1H-pyrazole ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
3-Amino-5-hydroxy-1H-pyrazole, 3-Amino-5-hydroxypyrazole, 5-Amino-3-pyrazolol, 3-Amino-1H-pyrazol-5(4H)-one, 3-Amino-2-pyrazolin-5-one, 5-Hydroxy-3-aminopyrazole, 5-Amino-2,4-dihydro-3H-pyrazol-3-one
A1589-25G
A1589-5G
TCIAA1589-25GEA
230
EUR
TCIAA1589-25G
TCIAA1589-5G
3-Amino-5-hydroxy-1H-pyrazole ≥98.0% (by HPLC, titration analysis)
3-Amino-5-hydroxy-1H-pyrazole
Formula:
C₃H₅N₃O MW: 99,09 g/mol |
Número MDL:
MFCD00022384 Núm. CAS: 6126-22-3 EINECS: 228-095-6 |
Resultados test especificaciones
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...