Sodium glycolate 97%

Proveedor: Thermo Fisher Scientific

351570250 351571000
ACRO351570250EA 30.1 EUR
ACRO351570250 ACRO351571000
Sodium glycolate 97%
Sodium glycolate
Formula: C₂H₃NaO₃
MW: 98.03 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00065358
Núm. CAS: 2836-32-0
EINECS: 220-624-9

Order Now

Resultados test especificaciones

Appearance White to cream Powder and/or lumps
Infrared spectrum Conforms
Titration with HClO4 96.0 to 104.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR