Sodium glycolate ≥97%
Proveedor: Thermo Fisher Scientific
A12341.0E
A12341.22
A12341.36
A12341.0EEA
673
EUR
A12341.0E
A12341.22
A12341.36
Sodium glycolate ≥97%
Sodium glycolate
Purity: 97%
CAS No.: 2836-32-0
Molecular Formula: C2H3NaO3
Formula Weight: 98.03
Physical Form: Lumpy powder
Appearance: White
Melting Point: 210-218°
MDL No.: MFCD00065358
Formula:
C₂H₃NaO₃ Pto. de fusión: 210…218 °C Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00065358 Núm. CAS: 2836-32-0 EINECS: 220-624-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...