4-Bromo-2-(trifluoromethoxy)anisole ≥98%
Proveedor: Thermo Fisher Scientific
|
Atención
|
H32645.03
H32645.06
H32645.03EA
43.3
EUR
H32645.03
H32645.06
4-Bromo-2-(trifluoromethoxy)anisole ≥98%
4-Bromo-2-(trifluoromethoxy)anisole
Purity: 98%
CAS No.: 853771-88-7
Molecular Formula: C8H6BrF3O2
Formula Weight: 271.04
MDL No.: MFCD09907950
Formula:
C₈H₆BrF₃O₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09907950 Núm. CAS: 853771-88-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...