4-Hydroxy-3-methoxybenzonitrile ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

H0620-25G H0620-5G
TCIAH0620-25GEA 131 EUR
TCIAH0620-25G TCIAH0620-5G
4-Hydroxy-3-methoxybenzonitrile ≥98.0% (by GC, titration analysis)
4-Hydroxy-3-methoxybenzonitrile
Formula: C₈H₇NO₂
MW: 149,15 g/mol
Número MDL: MFCD00001820
Núm. CAS: 4421-08-3
EINECS: 224-589-0
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab9688b11d3f9f30f87aaf62d6e32860922eaf64&vid=ab9688b11d3f9f30f87aaf62d6e32860922eaf64&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 86.0 to 89.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR