4-(Hydroxymethyl)phenylboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-(Hydroxymethyl)benzeneboronic acid
H1204-1G
H1204-5G
H1204-25G
TCIAH1204-1GEA
25
EUR
TCIAH1204-1G
TCIAH1204-5G
TCIAH1204-25G
4-(Hydroxymethyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(Hydroxymethyl)phenylboronic acid
Formula:
C₇H₉BO₃ MW: 151,96 g/mol |
Número MDL:
MFCD00792672 Núm. CAS: 59016-93-2 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (Neutralization titration) | 97.0 to 113.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...