Zinc selenite
Proveedor: Thermo Scientific
|
Peligro
|
12586.09
12586.22
12586.36
12586.A3
12586.09EA
18
EUR
12586.09
12586.22
12586.36
12586.A3
Zinc selenite
Zinc selenite
CAS No.: 13597-46-1
Molecular Formula: ZnSeO3
Formula Weight: 192.33
Physical Form: Powder
UN No.: 2630
MDL No.: MFCD00011297
Formula:
O₃SeZn MW: 192,33 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00011297 Núm. CAS: 13597-46-1 EINECS: 237-048-9 UN: 2630 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...