4,7-Dibromo-2,1,3-benzothiadiazole ≥98.0% (por GC)

Proveedor: TCI

D3842-1G D3842-25G D3842-5G
TCIAD3842-1GEA 23 EUR
TCIAD3842-1G TCIAD3842-25G TCIAD3842-5G
4,7-Dibromo-2,1,3-benzothiadiazole ≥98.0% (por GC)
4,7-Dibromo-2,1,3-benzothiadiazole
Formula: C₆H₂Br₂N₂S
MW: 293,97 g/mol
Número MDL: MFCD00658844
Núm. CAS: 15155-41-6
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae0d277c50bca11d4b97774b1a737e0eb51ee551&vid=ae0d277c50bca11d4b97774b1a737e0eb51ee551&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 187.0 to 191.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR