Diisopropyl phthalate ≥98.0% (por GC)

Proveedor: TCI
Atención

Sinónimos: Diisopropyl Phthalate

P0301-25ML P0301-500ML
TCIAP0301-25MLEA 37 EUR
TCIAP0301-25ML TCIAP0301-500ML
Diisopropyl phthalate ≥98.0% (por GC)
Diisopropyl phthalate
Formula: C₁₄H₁₈O₄
MW: 250,2903 g/mol
Número MDL: MFCD00053717
Núm. CAS: 605-45-8
EINECS: 210-086-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7fbbb858d23b8461edb60f4c887a4e743e54b8e4&vid=7fbbb858d23b8461edb60f4c887a4e743e54b8e4&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR