Rhodium(II) acetate dimer
Proveedor: TCI
Sinónimos:
Dirhodium tetraacetate, Tetrakis(acetato)dirhodium(II)
R0069-1G
R0069-100MG
TCIAR0069-1GEA
822
EUR
TCIAR0069-1G
TCIAR0069-100MG
Rhodium(II) acetate dimer
Rhodium(II) acetate dimer
Formula:
C₈H₁₂O₈Rh₂ MW: 441,987 g/mol |
Número MDL:
MFCD00003538 Núm. CAS: 15956-28-2 EINECS: 240-084-8 |
Resultados test especificaciones
Appearance | Green to Dark green powder to crystal |
Elemental analysis (Carbon) | 20.50 to 22.70 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...