4-Isopropylphenylboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-Isopropylbenzeneboronic acid
I0620-5G
I0620-1G
TCIAI0620-5GEA
81
EUR
TCIAI0620-5G
TCIAI0620-1G
4-Isopropylphenylboronic acid (contains varying amounts of Anhydride)
4-Isopropylphenylboronic acid
Formula:
C₉H₁₃BO₂ MW: 164,01 g/mol Pto. de fusión: 140°C |
Número MDL:
MFCD01074614 Núm. CAS: 16152-51-5 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 to 112.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...