D-(-)-α-(4-Hydroxyphenyl)glycine ≥99.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
D(-)-4-Hydroxyphenylglycine, H-D-Phg(4-OH)-OH, (R)-(-)-2-(4-Hydroxyphenyl)glycine, (-)-4-Hydroxy-D-phenylglycine, D-(-)-p-Hydroxyphenylglycine
H0758-25G
H0758-500G
H0758-100G
TCIAH0758-25GEA
17
EUR
TCIAH0758-25G
TCIAH0758-500G
TCIAH0758-100G
D-(-)-α-(4-Hydroxyphenyl)glycine ≥99.0% (by HPLC, titration analysis)
D-(-)-α-(4-Hydroxyphenyl)glycine
Formula:
C₈H₉NO₃ MW: 167,16 g/mol Pto. de fusión: 240°C |
Número MDL:
MFCD00004262 Núm. CAS: 22818-40-2 EINECS: 245-247-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (Nonaqueous Titration) | min. 99.0 % |
Specific rotation [a]20/D | -155.0 to -160.0 deg (C=1, 1mol/L HCl) |
Solubility in 1mol/L HCl | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...