2-Fluoroisonicotinic acid ≥97.0% (by GC, titration analysis)
Proveedor: TCI
Sinónimos:
2-Fluoropyridine-4-carboxylic acid
F0693-1G
F0693-5G
TCIAF0693-1GEA
50
EUR
TCIAF0693-1G
TCIAF0693-5G
2-Fluoroisonicotinic acid ≥97.0% (by GC, titration analysis)
2-Fluoroisonicotinic acid
Formula:
C₆H₄FNO₂ MW: 141,10 g/mol |
Número MDL:
MFCD02181194 Núm. CAS: 402-65-3 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...