2,4,6-Tribromobenzoic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

T0619-1G T0619-5G
TCIAT0619-1GEA 64 EUR
TCIAT0619-1G TCIAT0619-5G
2,4,6-Tribromobenzoic acid ≥98.0% (by GC, titration analysis)
2,4,6-Tribromobenzoic acid
Formula: C₇H₃Br₃O₂
MW: 358,8095 g/mol
Número MDL: MFCD00059882
Núm. CAS: 633-12-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a098131e8fd6d337d0c98101f7cddd528c15bbf&vid=3a098131e8fd6d337d0c98101f7cddd528c15bbf&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 192.0 to 196.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR