Isobutyl-4-hydroxybenzoate ≥99.0% (by titrimetric analysis) for biochemical research

Proveedor: TCI
Atención

Sinónimos: Isobutyl 4-hydroxybenzoate, Isobutylparaben

I0816-25G I0816-5G
TCIAI0816-25GEA 35 EUR
TCIAI0816-25G TCIAI0816-5G
Isobutyl-4-hydroxybenzoate ≥99.0% (by titrimetric analysis) for biochemical research
Isobutyl-4-hydroxybenzoate
Formula: C₁₁H₁₄O₃
Número MDL: MFCD00020167
Núm. CAS: 4247-02-3
EINECS: 224-208-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80dabe8eceddade9322e1acc1048bae0520e73af&vid=dbbf4566bc542613ec835a418d29349d4d2d5a05&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 99.0 %
Melting point 75.0 to 77.0 deg-C
Protein impurity test (SDS-PAGE) to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR