3-Nitrosalicylic acid ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
N0251-25G
N0251-5G
TCIAN0251-25GEA
175
EUR
TCIAN0251-25G
TCIAN0251-5G
3-Nitrosalicylic acid ≥98.0% (by GC, titration analysis)
3-Nitrosalicylic acid
Formula:
C₇H₅NO₅ |
Número MDL:
MFCD00024240 Núm. CAS: 85-38-1 Índice Merck: 14,06631 |
|
Resultados test especificaciones
Appearance | White to Light yellow to Green powder to crystaline |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 145.0 to 150.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...