Ácido 3,5,6-triclorosalicílico ≥97.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
3,5,6-Trichlorosalicylic Acid
T1406-25G
T1406-500G
TCIAT1406-25GEA
30
EUR
TCIAT1406-25G
TCIAT1406-500G
Ácido 3,5,6-triclorosalicílico ≥97.0% (by GC, titration analysis)
Ácido 3,5,6-triclorosalicílico
Formula:
C₇H₃Cl₃O₃ MW: 241,46 g/mol |
Número MDL:
MFCD00075245 Núm. CAS: 40932-60-3 EINECS: 255-144-9 |
Resultados test especificaciones
Appearance | White to Gray to Red powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...