2-Chloro-4-fluoro-5-nitrobenzoic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

C2446-5G C2446-25G
TCIAC2446-5GEA 65 EUR
TCIAC2446-5G TCIAC2446-25G
2-Chloro-4-fluoro-5-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
2-Chloro-4-fluoro-5-nitrobenzoic acid
Formula: C₇H₃ClFNO
MW: 219,55 g/mol
Número MDL: MFCD04115715
Núm. CAS: 114776-15-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fa4be4e811040f5d184948464c232f71458a0a7&vid=6fa4be4e811040f5d184948464c232f71458a0a7&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 152.0 to 156.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR