4,4'-Dibromo-trans-stilbene ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
(E)-1,2-Bis(4-bromophenyl)ethene
D3043-1G
D3043-250MG
D3043-5G
TCIAD3043-1GEA
34
EUR
TCIAD3043-1G
TCIAD3043-250MG
TCIAD3043-5G
4,4'-Dibromo-trans-stilbene ≥98.0% (por GC)
4,4'-Dibromo-trans-stilbene
Formula:
C₁₄H₁₀Br₂ MW: 338,0372 g/mol |
Número MDL:
MFCD00448009 Núm. CAS: 18869-30-2 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 210.0 to 215.0 deg-C |
Solubility in hot Toluene | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...