DL-Glutamic acid ≥92.0% (by HPLC)
Proveedor: TCI
G0058-25G
G0058-500G
TCIAG0058-25GEA
26
EUR
TCIAG0058-25G
TCIAG0058-500G
DL-Glutamic acid ≥92.0% (by HPLC)
DL-Glutamic acid
Formula:
C₅H₉NO₄ MW: 147,13 g/mol |
Número MDL:
MFCD00063113 Núm. CAS: 617-65-2 EINECS: 210-522-2 Índice Merck: 14,04469 |
Resultados test especificaciones
Appearance | White powder to crystal |
Purity (HPLC) (CAD) | min. 92.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...