4-Methylphthalic acid ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

M0560-25G M0560-500G
TCIAM0560-25GEA 43 EUR
TCIAM0560-25G TCIAM0560-500G
4-Methylphthalic acid ≥98.0% (by HPLC, titration analysis)
4-Methylphthalic acid
Formula: C₉H₈O₄
Número MDL: MFCD00041946
Núm. CAS: 4316-23-8
EINECS: 224-333-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8762d0d7345da32d30995e6c9050d3e2784c2d57&vid=8762d0d7345da32d30995e6c9050d3e2784c2d57&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR