Potassium selenite
Proveedor: Thermo Fisher Scientific
|
Peligro
|
39436.14
39436.22
39436.14EA
125
EUR
39436.14
39436.22
Potassium selenite
Potassium selenite
CAS No.: 10431-47-7
Molecular Formula: K2SeO3
Formula Weight: 205.16
Physical Form: Powder
Density: 2.851
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 875° dec.
UN No.: 2630
MDL No.: MFCD00049663
Formula:
K₂O₃Se Pto. de fusión: 875 °C (desc.) Densidad: 2,851 Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00049663 Núm. CAS: 10431-47-7 EINECS: 233-909-8 UN: 2630 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...