3-(4-Methoxybenzoyl)propionic acid ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-(4-Methoxyphenyl)-4-oxobutanoic acid
M1383-25G
M1383-5G
TCIAM1383-25GEA
130
EUR
TCIAM1383-25G
TCIAM1383-5G
3-(4-Methoxybenzoyl)propionic acid ≥98.0% (by HPLC, titration analysis)
3-(4-Methoxybenzoyl)propionic acid
Formula:
C₁₁H₁₂O₄ MW: 208,21 g/mol |
Número MDL:
MFCD00002795 Núm. CAS: 3153-44-4 EINECS: 221-593-4 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 148.0 to 152.0 deg-C |
Solubility in hot Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...