Methyl (4-fluorophenyl)acetate ≥98%

Proveedor: Thermo Fisher Scientific

H32236.06 H32236.14
H32236.06EA 48.4 EUR
H32236.06 H32236.14
Methyl (4-fluorophenyl)acetate ≥98%
Methyl (4-fluorophenyl)acetate

Purity: 98% CAS No.: 34837-84-8 Molecular Formula: C9H9FO2 Formula Weight: 168.17 MDL No.: MFCD00800608

Formula: C₉H₉FO₂
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00800608
Núm. CAS: 34837-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR