Ácido 3-Cianofenilborónico (contains varying amounts of Anhydride)

Proveedor: TCI
Peligro

Sinónimos: (3-Cyanophenyl)metaboric acid, Dihydroxy(3-cyanophenyl)borane, 3-Cyanobenzeneboronic acid

C2031-25G C2031-5G
TCIAC2031-25GEA 170 EUR
TCIAC2031-25G TCIAC2031-5G
Ácido 3-Cianofenilborónico (contains varying amounts of Anhydride)
Ácido 3-Cianofenilborónico
Formula: C₇H₆BNO₂
MW: 146.94 g/mol
Número MDL: MFCD01318967
Núm. CAS: 150255-96-2
UN: 3439
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=931a4eb046f1a4a1c258352214fe7fbab39e35f2&vid=931a4eb046f1a4a1c258352214fe7fbab39e35f2&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR