Ácido (α-α-α-trifluoro-4-tolil)acetico ≥97.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: Ácido (4-(trifluorometil)fenil)acético

T1783-1G T1783-25G T1783-5G
TCIAT1783-1GEA 23 EUR
TCIAT1783-1G TCIAT1783-25G TCIAT1783-5G
Ácido (α-α-α-trifluoro-4-tolil)acetico ≥97.0% (by titrimetric analysis)
Ácido (α-α-α-trifluoro-4-tolil)acetico
Formula: (C₆H₄(CF₃))CH₂COOH
Número MDL: MFCD00004352
Núm. CAS: 32857-62-8
EINECS: 251-263-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6799ddd765529c97c88ae374f510af358a291c61&vid=6799ddd765529c97c88ae374f510af358a291c61&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 97.0 %
Melting point 82.0 to 85.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR