Dicloruro de benceno-1,2-disulfonilo ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Peligro
|
B1128-1G
B1128-25G
B1128-5G
TCIAB1128-1GEA
88
EUR
TCIAB1128-1G
TCIAB1128-25G
TCIAB1128-5G
Dicloruro de benceno-1,2-disulfonilo ≥98.0% (by titrimetric analysis)
Dicloruro de benceno-1,2-disulfonilo
Formula:
C₆H₄Cl₂O₄S₂ |
Número MDL:
MFCD00059836 Núm. CAS: 6461-76-3 UN: 3261 ADR: 8,III |
|
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 144.0 to 147.0 deg-C |
Solubility in hot Toluene | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...