Bis(2,4-pentanodionato-O,O')níquel (II) hidrato ≥85.0% (by titrimetric analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
Acetylacetone nickel(II) salt hydrate, Nickel (II) bis(2,4-pentanedionato-O,O') hydrate, Bis(2,4-pentanedionato-O,O')nickel (II) hydrate, Nickel (II) 2,4-pentanedionate hydrate, Bis(2,4-pentanedionato)nickel(II) hydrate
N0096-25G
N0096-500G
N0096-100G
TCIAN0096-25GEA
44
EUR
TCIAN0096-25G
TCIAN0096-500G
TCIAN0096-100G
Bis(2,4-pentanodionato-O,O')níquel (II) hidrato ≥85.0% (by titrimetric analysis)
Bis(2,4-pentanodionato-O,O')níquel (II) hidrato
Formula:
C₁₀H₁₄NiO₄·xH₂O MW: 256.91 g/mol (anhy) |
Número MDL:
MFCD00149058 Núm. CAS: 120156-44-7 EINECS: 221-875-7 Índice Merck: 14,06500 |
|
Resultados test especificaciones
Appearance | White to Dark green to Dark purple powder to crystal |
Purity (Chelometric Titration) | min. 85.0 % |
Solubility in hot Methanol | very faint turbidity |
Water | max. 14.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...