L(+)-Ramnosa monohidrato ≥98.0% (by HPLC)
Proveedor: TCI
Sinónimos:
6-Desoxi-L(+)-manosa monohidrato
R0013-25G
R0013-5G
TCIAR0013-25GEA
56
EUR
TCIAR0013-25G
TCIAR0013-5G
L(+)-Ramnosa monohidrato ≥98.0% (by HPLC)
L(+)-Ramnosa monohidrato
Formula:
C₆H₁₂O₅·1H₂O MW: 182.17 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00149363 Núm. CAS: 10030-85-0 EINECS: 222-793-4 Índice Merck: 14,08172 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 90.0 to 95.0 deg-C |
Specific rotation [a]20/D | +8.0 to +8.5 deg (C=10, H2O)[included NH3 aq. (2+3)] |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...