(R)-(-)-Ethyl mandelate ≥98.0% (por GC)

Proveedor: TCI

Sinónimos: D(-)-Glicolato de etilo

M1344-25G M1344-5G
TCIAM1344-25GEA 139 EUR
TCIAM1344-25G TCIAM1344-5G
(R)-(-)-Ethyl mandelate ≥98.0% (por GC)
(R)-(-)-Ethyl mandelate
Formula: C₁₀H₁₂O₃
MW: 180,2 g/mol
Número MDL: MFCD00064248
Núm. CAS: 10606-72-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=382e75b7e9d81b326eb1e08ed26aedec4502fcb6&vid=382e75b7e9d81b326eb1e08ed26aedec4502fcb6&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Optical purity (LC) min. 95.0 ee%
Freezing point 26.0 to 31.0 deg-C
Specific rotation [a]20/D -130 to -145 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR