Ácido 4-acetil-fenil-borónico (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
(4-Acetylphenyl)metaboric acid, 4-Acetylbenzeneboronic acid, Dihydroxy(4-acetylphenyl)borane
A1907-1G
A1907-5G
TCIAA1907-1GEA
16
EUR
TCIAA1907-1G
TCIAA1907-5G
Ácido 4-acetil-fenil-borónico (contains varying amounts of Anhydride)
Ácido 4-acetil-fenil-borónico
Formula:
C₈H₉BO₃ |
Número MDL:
MFCD01074667 Núm. CAS: 149104-90-5 |
|
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to crystal |
Purity (Neutralization titration) | 97.0 to 112.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...