Ácido 5-Cloro-2-tiofenoborónico (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
C2066-1G
C2066-5G
TCIAC2066-1GEA
34
EUR
TCIAC2066-1G
TCIAC2066-5G
Ácido 5-Cloro-2-tiofenoborónico (contains varying amounts of Anhydride)
Ácido 5-Cloro-2-tiofenoborónico
Formula:
C₄H₄BClO₂S |
Número MDL:
MFCD00236030 Núm. CAS: 162607-18-3 |
|
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 to 112.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...