Bis(2,4-pentanodionato-O,O')magnesio ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Magnesium bis(2,4-pentanedionato-O,O'), Bis(2,4-pentanedionato-O,O')magnesium, Magnesiumacetylacetonate, Magnesium 2,4-pentanedionate
M0001-25G
M0001-5G
TCIAM0001-25GEA
124
EUR
TCIAM0001-25G
TCIAM0001-5G
Bis(2,4-pentanodionato-O,O')magnesio ≥98.0% (by titrimetric analysis)
Bis(2,4-pentanodionato-O,O')magnesio
Formula:
C₁₀H₁₄MgO₄ MW: 222.52 g/mol |
Número MDL:
MFCD00013497 Núm. CAS: 14024-56-7 EINECS: 237-857-7 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Chelometric Titration) | min. 98.0 % |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...