D-alo-Treonina ≥99.0% (by HPLC, titration analysis)
Proveedor: TCI
A1518-1G
A1518-100MG
TCIAA1518-1GEA
153
EUR
TCIAA1518-1G
TCIAA1518-100MG
D-alo-Treonina ≥99.0% (by HPLC, titration analysis)
D-alo-Treonina
Formula:
C₄H₉NO₃ |
Número MDL:
MFCD00004526 Núm. CAS: 24830-94-2 EINECS: 246-488-0 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (Nonaqueous Titration) | min. 99.0 % |
Specific rotation [a]20/D | -9.0 to -11.0 deg (C=5, H2O) |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...