L-alo-Treonina ≥99.0% (by HPLC)

Proveedor: TCI

Sinónimos: L(+)-alo-Treonina, H-allo-Thr-OH

A1519-1G A1519-100MG
TCIAA1519-1GEA 138 EUR
TCIAA1519-1G TCIAA1519-100MG
L-alo-Treonina ≥99.0% (by HPLC)
L-alo-Treonina
Formula: C₄H₉NO₃
MW: 119,12 g/mol
Número MDL: MFCD00064268
Núm. CAS: 28954-12-3
EINECS: 249-327-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7eb0dbb49f362c8d3da4b83f1146a45222ed954&vid=a7eb0dbb49f362c8d3da4b83f1146a45222ed954&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +9.0 to +11.0 deg (C=5, H2O)
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR