Ácido glioxilico ∼50% ∼9 mol/L in water

Proveedor: TCI
Peligro

Sinónimos: 2-Oxoacetic acid

G0366-25ML G0366-500ML
TCIAG0366-25MLEA 16 EUR
TCIAG0366-25ML TCIAG0366-500ML
Ácido glioxilico ∼50% ∼9 mol/L in water
Ácido glioxilico
Formula: C₂H₂O₃
MW: 74,04 g/mol
Número MDL: MFCD00006958
Núm. CAS: 298-12-4
EINECS: 206-058-5
UN: 3265
ADR: 8,III
Índice Merck: 14,04511
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2fb5238013e2c9070938782ca61c4713047711a&vid=bcdfec2e514e4d8308e67f8096a4e00111536140&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Colorless to Light orange to Yellow clear liquid
Concentration (Neutralization titration) 50.0 to 55.0 w/w% (calcd. on exclusive of Oxalic acid)
Oxalic Acid max. 4.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR