Cloruro de 1-(2-cloroetil)piperidinio ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Peligro
|
C1073-25G
C1073-500G
TCIAC1073-25GEA
34
EUR
TCIAC1073-25G
TCIAC1073-500G
Cloruro de 1-(2-cloroetil)piperidinio ≥98.0% (by titrimetric analysis)
Cloruro de 1-(2-cloroetil)piperidinio
Formula:
C₇H₁₅Cl₂N |
Número MDL:
MFCD00012837 Núm. CAS: 2008-75-5 EINECS: 217-920-5 UN: 2928 ADR: 6.1,II |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...