4-Acetamido-2,2,6,6-tetrametilpiperidinooxi (4-acetamido-TEMPO) ≥98.0% (por GC) catalyst for oxidation free radical
Proveedor: TCI
|
Atención
|
Sinónimos:
4-Acetamido-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Acetamido-2,2,6,6-tetramethylpiperidine 1-oxyl, 4-Acetamido-2,2,6,6-Tetramethyl-N-Piperidinyloxy
A1348-25G
A1348-5G
TCIAA1348-25GEA
180
EUR
TCIAA1348-25G
TCIAA1348-5G
4-Acetamido-2,2,6,6-tetrametilpiperidinooxi (4-acetamido-TEMPO) ≥98.0% (por GC) catalyst for oxidation free radical
4-Acetamido-2,2,6,6-tetrametilpiperidinooxi (4-acetamido-TEMPO)
Formula:
C₁₁H₂₁N₂O₂ MW: 213,3 g/mol |
Número MDL:
MFCD00043593 Núm. CAS: 14691-89-5 EINECS: 423-840-3 |
Resultados test especificaciones
Appearance | Light yellow to Brown powder to crystaline |
Purity (GC) | min. 98.0 % |
Purity (Iodometric Titration) | min. 96.0 % |
Melting point | 143.0 to 147.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...