Cloroformiato de colest-5-eno-3β-ilo ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

C0694-25G C0694-100G
TCIAC0694-25GEA 75 EUR
TCIAC0694-25G TCIAC0694-100G
Cloroformiato de colest-5-eno-3β-ilo ≥98.0% (by titrimetric analysis)
Cloroformiato de colest-5-eno-3β-ilo
Formula: C₂₈H₄₅ClO₂
MW: 449,12 g/mol
Número MDL: MFCD00003633
Núm. CAS: 7144-08-3
EINECS: 230-447-9
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6dc07c62fad8af7df3c1e8c69f923c34170a31fa&vid=6dc07c62fad8af7df3c1e8c69f923c34170a31fa&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Melting point 116.0 to 120.0 deg-C
Specific rotation [a]20/D -21.0 to -25.0 deg (C=2, THF)
Solubility in THF almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR