Dicloruro de 4-hidroxi-m-fenilendiamonio ≥97.0% (by total nitrogen and titration analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
4-Hydroxy-1,3-Phenylenediammonium Dichloride, 2,4-Diaminophenol Dihydrochloride, Amidol
D0080-25G
D0080-500G
TCIAD0080-25GEA
24
EUR
TCIAD0080-25G
TCIAD0080-500G
Dicloruro de 4-hidroxi-m-fenilendiamonio ≥97.0% (by total nitrogen and titration analysis)
Dicloruro de 4-hidroxi-m-fenilendiamonio
Formula:
C₆H₁₀Cl₂N₂O Pto. de fusión: 205°C |
Número MDL:
MFCD00012979 Núm. CAS: 137-09-7 EINECS: 205-279-4 UN: 2811 ADR: 6.1,III Índice Merck: 14,02983 |
Resultados test especificaciones
Appearance | White to Amber to Dark green powder to crystal |
Purity (Argentmetric Titration) | min. 97.0 % (calcd.on dried substance) |
Purity (with Total Nitrogen) | min. 98.0 % (calcd.on dried substance) |
Drying loss | max. 5.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...