Purpura de m-cresol

Proveedor: TCI

M0074-1G M0074-25G
TCIAM0074-1GEA 16 EUR
TCIAM0074-1G TCIAM0074-25G
Purpura de m-cresol
Purpura de m-cresol
Formula: C₂₁H₁₈O₅S
Número MDL: MFCD00005871
Núm. CAS: 2303-01-7
EINECS: 218-960-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=128c7f9762c89b34e8de4b1246a2030d7225e05d&vid=128c7f9762c89b34e8de4b1246a2030d7225e05d&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Yellow to Brown to Dark green powder to crystaline
Transition interval (pH) (red)1.2 ~ 2.8 (yellow), (yellow)7.4 ~ 9.0 (purple)
Lambda max. 432.0 to 436.0 nm
Absorbance min. 0.360 (10 mg/L, pH 7.0, 432.0 to 436.0 nm)

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR