N-Benciloxicarbonilglicina ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
N-Carbobenzyloxyglycine, N-Benzyloxycarbonylglycine, Z-Gly-OH, 2-(((Benzyloxy)carbonyl)amino)acetic acid
C0575-25G
C0575-500G
TCIAC0575-25GEA
16
EUR
TCIAC0575-25G
TCIAC0575-500G
N-Benciloxicarbonilglicina ≥98.0% (by HPLC, titration analysis)
N-Benciloxicarbonilglicina
Formula:
C₁₀H₁₁NO₄ MW: 209,2 g/mol |
Número MDL:
MFCD00002691 Núm. CAS: 1138-80-3 EINECS: 214-516-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 118.0 to 122.0 deg-C |
Solubility in hot Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...