Calcein (mixture of isomers) for fluorometric determination of Ca

Proveedor: TCI

Sinónimos: Fluorexon (mixture of isomers), Fluorescein Bis(methyliminodiacetic Acid) (mixture of isomers), N,N'-[(3',6'-Dihydroxy-3-oxospiro[isobenzofuran-1(3H),9'-[9H]xanthene]-ar',ar'-diyl)bis(methylene)]bis[N-(carboxymethyl)glycine], Bis[N,N-bis(carboxymethyl)aminomethyl]fluorescein (mixture of isomers), Calcein

C0004-1G C0004-5G
TCIAC0004-1GEA 79 EUR
TCIAC0004-1G TCIAC0004-5G
Calcein (mixture of isomers) for fluorometric determination of Ca
Calcein (mixture of isomers)
Formula: C₃₀H₂₆N₂O₁₃
MW: 622,53 g/mol
Número MDL: MFCD00005049
Núm. CAS: 154071-48-4
EINECS: 215-957-1

Order Now

Resultados test especificaciones

Appearance Orange to Brown powder to crystaline
Drying loss max. 6.0 %
Ignition residue (Sulfate) max. 1.0 %
Fluorescein max. 0.05 %
Sensitiveness to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR