Sulfato de p-fenilendiamina ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Peligro
|
P0175-500G
P0175-25G
TCIAP0175-500GEA
58
EUR
TCIAP0175-500G
TCIAP0175-25G
Sulfato de p-fenilendiamina ≥98.0% (by titrimetric analysis)
Sulfato de p-fenilendiamina
Formula:
C₆H₁₀N₂O₄S |
Número MDL:
MFCD00035510 Núm. CAS: 16245-77-5 EINECS: 240-357-1 UN: 2811 ADR: 6.1,III |
|
Resultados test especificaciones
Appearance | White to Amber to Dark purple powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...