(±)-2-Amino-2-(4-chlorophenyl)acetic acid ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: DL-2-(4-Chlorophenyl)glycine, H-DL-Phg(4-Cl)-OH, DL-alpha-Amino-4-chlorophenylacetic acid, 4-Chloro-DL-2-phenylglycine, 4-Chloro-DL-phenylglycine, 2-Amino-2-(4-chlorophenyl)acetic acid

C1401-1G C1401-5G C1401-25G
TCIAC1401-1GEA 16 EUR
TCIAC1401-1G TCIAC1401-5G TCIAC1401-25G
(±)-2-Amino-2-(4-chlorophenyl)acetic acid ≥98.0% (by titrimetric analysis)
(±)-2-Amino-2-(4-chlorophenyl)acetic acid
Formula: C₈H₈ClNO₂
MW: 185,61 g/mol
Número MDL: MFCD00049328
Núm. CAS: 6212-33-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f16eb39531f543ee42b24468ee5f350aaa694968&vid=f16eb39531f543ee42b24468ee5f350aaa694968&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in 0.1mol/L NaOH almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR